Learn More
Thermo Scientific™ Oxoid™ Prepared Reinforced Clostridial Bottled Media
Click to view available options
:
Volume: 100mL
Descrizione
Oxoid™ Prepared Reinforced Clostridial Bottled Media, 100 mL
Specifica
Specifica
| Product Type | Microbiology Media |