Learn More
Thermo Scientific™ Oxoid™ Prepared Tryptone Water Bijou Bottled Media
Codice prodotto. 12736969
Click to view available options
:
Volume: 5mL
Codice prodotto. 12736969
Marca: Thermo Scientific™ BO0383C
Tryptone Water is a liquid medium recommended for use in qualitative procedures for detection of Escherichia coli in food and water samples based on indole production.