missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Skirrow Campylobacter Selective Supplement
Click to view available options
Resa:
Ciascun flacone per 500 ml di terreno
Each vial for 2L medium
Dimensione della confezione:
pezzi 10
Description
Oxoid Campylobacter Selective Supplement, Skirrow is used for preparation of Skirrow Selective Medium for the isolation of Campylobacter spp. at 42°C.
Add to Blood Agar Base No. 2, Part No. CM0271B or Columbia Blood Agar Base, Part No. CM0331B
Specifications
Specifications
| Descrizione | Supplemento selettivo Campylobacter, Skirrow |
| Forma | Liofilizzato |
| Product Type | Selective Supplement |
| Quantità | 10/confezione |
| Resa | Ciascun flacone per 500 ml di terreno |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction