Learn More
Thermo Scientific™ Oxoid™ Skirrow Campylobacter Selective Supplement
Codice prodotto. p-6081167
Click to view available options
Resa:
Ciascun flacone per 500 ml di terreno
Each vial for 2L medium
Dimensione della confezione:
pezzi 10
Codice prodotto. 11494149
Marca: Thermo Scientific™ SR0069E
Oxoid Campylobacter Selective Supplement, Skirrow prepares Skirrow Selective Medium for isolation of Campylobacter spp. at 42°C.
Oxoid Campylobacter Selective Supplement, Skirrow is used for preparation of Skirrow Selective Medium for the isolation of Campylobacter spp. at 42°C.
Add to Blood Agar Base No. 2, Part No. CM0271B or Columbia Blood Agar Base, Part No. CM0331B
TRUSTED_SUSTAINABILITY
Specifica
Descrizione Supplemento selettivo Campylobacter, Skirrow
Forma Liofilizzato
Product Type Selective Supplement
Quantità 10/confezione
Resa Ciascun flacone per 500 ml di terreno