Learn More
Thermo Scientific™ Oxoid™ STAA Selective Supplement
Click to view available options
Quantità:
10/Pk.
Dimensione della confezione:
pezzi 10
Descrizione
Oxoid STAA Selective Supplement is used for the isolation of Brochothrix thermosphacta from food samples.
- Add to STAA Agar Base, Part No. CM0881B
- Each vial supplements 500mL of medium
Specifica
Specifica
| Descrizione | STAA Selective Supplement |
| Forma | Freeze-dried |
| Product Type | Selective Supplement |
| Quantità | 10/Pk. |
| Resa | Each vial for 500mL medium |