Learn More
Thermo Scientific™ Oxoid™ Tobramycin Antimicrobial Susceptibility discs
Click to view available options
Concentrazione:
10 µg
30 μg
Dimensione della confezione:
pezzi 250
Descrizione
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Tobramycin Antimicrobial Susceptibility discs.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
- Inoculum level
- Presence of contamination
- Resistant mutants
- Beta-lactamase activity
- Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifica
Specifica
| Agenti antimicrobici | Tobramycin |
| Certificazione/Conformità | Conforme alle linee guida CLSI, DIN, BSAC, EUCAST, SFM |
| Concentrazione | 10 µg |
| Descrizione | Dischi di sensibilità antimicrobica Oxoid con tobramicina |
| Codice antibiotico | TOB |
| Product Type | Antimicrobial Susceptibility Discs |
| Test Type | Antimicrobial Susceptibility |
| N. per confezione | 5 Cartridges/Pk. |
| Tipo di imballaggio | 5 cartucce/confezione |
| Quantità | 50 dischi/cartuccia |
| Vedi altri risultati |