Learn More
Thermo Scientific™ Oxoid™ VCNT Selective Supplement
Click to view available options
Resa:
Each vial for 2L medium
Each vial for 500mL medium
Dimensione della confezione:
pezzi 10
Descrizione
Oxoid VCNT Selective Supplement is used for the preparation of Thayer Martin Medium for the isolation of pathogenic Neisseria spp.
- Add to GC Agar Base, Part No. CM0367B
Specifica
Specifica
| Descrizione | VCNT Selective Supplement |
| Forma | Freeze-dried |
| Product Type | Selective Supplement |
| Quantità | 10/Pk. |
| Resa | Each vial for 500mL medium |