Learn More
P-cadherin Rabbit anti-Human, Biotin, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech BIOTIN-13773-100UL
Descrizione
The P-cadherin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human P-cadherin. This antibody recognizes human, mouse, rat antigen. The P-cadherin antibody has been validated for the following applications: IHC analysis. This antibody is biotin.
Specifica
| P-cadherin | |
| Polyclonal | |
| Biotin | |
| 100 μL | |
| 1001 | |
| IgG |
| Immunohistochemistry | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |