missing translation for 'onlineSavingsMsg'
Learn More

p-Chloranil, 97%

Codice prodotto. 11425593
Click to view available options
Quantità:
25 g
100 g
500 g
Dimensione della confezione:
100g
25g
500g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 11425593

Marca: Thermo Scientific Alfa Aesar A13495.14

Richiesta di formato massivo o personalizzato

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 118-75-2 | C6Cl4O2 | 245.86 g/mol

p-Chloranil is used as a dye intermediate, oxidizing agent, vulcanizing agent and dehydrogenation reagent. It is also used to make chloranil electrodes for pH measurements. Further, it serves as a hydrogen acceptor and used for the aromatization reactions such as conversion of cyclohexadienes to the benzene derivatives. It is used to test the secondary amine. In addition to this, it is employed as a precursor to prepare diaziquone, which is used as a cancer chemotherapeutic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Chloranil is used as a dye intermediate, oxidizing agent, vulcanizing agent and dehydrogenation reagent. It is also used to make chloranil electrodes for pH measurements. Further, it serves as a hydrogen acceptor and used for the aromatization reactions such as conversion of cyclohexadienes to the benzene derivatives. It is used to test the secondary amine. In addition to this, it is employed as a precursor to prepare diaziquone, which is used as a cancer chemotherapeutic agent.

Solubility
Soluble in ether and acetone. Slightly soluble in chloroform, dimethylformamide, solvent naphtha, benzene, carbon disulfide and carbon tetrachloride. Insoluble in water, cold petroleum ether, methanol, dibutyl phthalate and cold alcohol.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 118-75-2
Formula molecolare C6Cl4O2
Molecular Weight (g/mol) 245.86
Numero MDL MFCD00001594
InChI Key UGNWTBMOAKPKBL-UHFFFAOYSA-N
Sinonimo chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone
PubChem CID 8371
ChEBI CHEBI:36703
IUPAC Name 2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione
SMILES ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O

Specifica

Materiale o nome chimico p-Chloranil
Punto di fusione ∼290°C (sublimation)
Densità 1.97
Quantità 25 g
Numero UN UN3077
Beilstein 393006
Indice di Merck 14,2078
Informazioni di solubilità Soluble in ether and acetone. Slightly soluble in chloroform,dimethylformamide,solvent naphtha,benzene,carbon disulfide and carbon tetrachloride. Insoluble in water,cold petroleum ether,methanol,dibutyl phthalate and cold alcohol.
Peso formulazione 245.88
Percent Purity 97%
compliance-icons compliance-icons
Identificativo prodotto
  • p-Chloranil
Avvertenza
  • Pericolo
Categoria di pericolo
  • PERICOLO A LUNGO TERMINE PER L'AMBIENTE ACQUATICO Cronico 1
  • PERICOLO ACUTO PER L'AMBIENTE ACQUATICO Acuto 1
  • Lesione oculare grave/irritazione oculare Categoria 2
  • Corrosione/irritazione della pelle Categoria 2
Indicazioni di pericolo
  • H315-Provoca irritazione cutanea.
  • H319-Provoca grave irritazione oculare.
  • H400-Molto tossico per gli organismi acquatici.
  • H410-Molto tossico per gli organismi acquatici con effetti di lunga durata.
Consigli di prudenza
  • P264-Lavare accuratamente dopo l'uso.
  • P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
  • P302+P352-IN CASO DI CONTATTO CON LA PELLE: lavare abbondantemente con acqua/.
  • P305+P351+P338-IN CASO DI CONTATTO CON GLI OCCHI: sciacquare accuratamente per parecchi minuti. Togliere le eventuali lenti a contatto se è agevole farlo. Continuare a sciacquare.
  • P332+P313-In caso di irritazione della pelle: consultare un medico.
  • P337+P313-Se l'irritazione degli occhi persiste, consultare un medico.
  • P362+P364-Togliere tutti gli indumenti contaminati e lavarli prima di indossarli nuovamente.
  • P403-Conservare in luogo ben ventilato.
  • P501b-Smaltire il prodotto/recipiente in conformità con le disposizioni locali/regionali/nazionali/internazionali.
Informazioni supplementari
  • MIXTURE LIST-Contiene: 2,3,5,6-Tetrachloro-2,5-cyclohexadiene-1,4-dione

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato