missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ P-Selectin Monoclonal Antibody (Psel.KO.2.7), PE

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA181849
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti human CD62P, clone Psel.KO.2.7, recognizes human P-Selectin.
P-selectin (Selectin P, GMP-140, LECAM3, CD62 antigen-like family member) is a 140 kDa type 1 transmembrane glycoprotein, and is one of the most commonly studied proteins that regulate cell rolling. P-selectin is stored in the Weibel-Palade bodies of endothelial cells, as well as in a-granules of platelets. From there, it can be rapidly brought to the cell surface after exposure to thrombin, histamine, complement 5a, Ca21 ionophores, or adenosine diphosphate. P-selectin protein redistributes to the plasma membrane during platelet activation and degranulation and mediates the interaction of activated endothelial cells or platelets with leukocytes. P-selectin plays an important role in adhesive processes between leucocytes and endothelial cells, and is a calcium dependent receptor that binds to sialylated forms of Lewis blood group carbohydrate antigens found on neutrophils and monocytes. P-selectin is constitutively expressed in inflammation and contributes to atherogenesis, thrombosis and tissue destruction. Clinically, P-selectin is used to distinguish heparin induced thrombocytopenia with and without thrombosis.
Specifica
| P-Selectin | |
| Monoclonal | |
| Conc. Not Determined | |
| PBS with 1% BSA, 5% sucrose and 0.09% sodium azide; pH 7.4 | |
| P16109, P42201, P98106, P98109, Q01102 | |
| SELP | |
| P-selectin transfected 300.19 cells. | |
| 100 Tests | |
| Primary | |
| Human, Mouse, Rat, Bovine, Sheep, Feline, Horse, Goat | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| Psel.KO.2.7 | |
| PE | |
| SELP | |
| CD62; CD62 antigen-like family member P; CD62P; CD62P antigen; CD62P; CD62 homolog; FLJ45155; GMP140; GMP-140; GMRP; Granule membrane protein 140; granule membrane protein 140kDa; granulocyte membrane protein; Grmp; LECAM3; leukocyte-endothelial cell adhesion molecule 3; PADGEM; Platelet activation dependent granule-external membrane protein; platelet alpha-granule membrane protein; PSEL; PSELECT; P-selectin; P-selectin precursor; RP1-86F14.2; selectin P; selectin P (granule membrane protein 140kDa, antigen CD62); selectin, platelet; Selp | |
| Mouse | |
| Protein A | |
| RUO | |
| 100009695, 101083565, 102170076, 20344, 25651, 281486, 443305, 6403 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Lyophilized |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto