missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P-selectin Mouse anti-Human, Clone: 3G2C7, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60322-1-IG-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
P-Selectin, also called CD62, GRMP, GMP140 is a 140-kD adhesion molecule, expressed at the surface of activated cells, that mediates the interaction of activated endothelial cells or platelets with leukocytes. The P-Selectin gene is mapped on 1q24.2. P-selectin plays an essential role in the initial recruitment of leukocytes (white blood cells) to the site of injury during inflammation. P-selectin is also very important in the recruitment and aggregation of platelets at areas of vascular injury. In quiescent platelet, P-selectin is located on the inner wall of alpha-granules. Sorting nexin-17 interacts with the cytosolic domain of P-selectin and suggested that SNX17 may function in the intracellular trafficking of P-selectin. P-selectin expression was found to be significantly and positively correlated with carotid artery stiffness and wall thickness, and the percentage of P-selectin-positive platelets was significantly higher in patients with carotid plaque.Specifica
| P-selectin | |
| Monoclonal | |
| 1.08 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P16109 | |
| SELP | |
| P-selectin Fusion Protein Ag4124 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 3G2C7 | |
| Unconjugated | |
| SELP | |
| CD62, CD62P, GMP 140, GMP140, GMRP, Granule membrane protein 140, GRMP, LECAM3, P selectin, PADGEM, PSEL, P-selectin, SELP | |
| Mouse | |
| Protein A | |
| RUO | |
| 6403 | |
| -20°C | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto