Learn More
Cole-Parmer™ RE3022C Diaphragm Vacuum Pump
Click to view available options
Tipo di pompa:
Pompa da vuoto
Dimensione della confezione:
Pezzo
Descrizione
- Solid PTFE and FFKM construction resists most solvents
- Regulate vacuum level using adjustment knob
- Integral catch pots prevent liquid ingress
- 3 year warranty
- Previous part number RE3022C
Use this compact diaphragm vacuum pump for chemical laboratory vacuum generation, especially well suited to rotary evaporation. Vacuum regulator with gauge and adjustment knob allows you to create ideal vacuum conditions in the lab. Oilless pump with integrated moisture protection is low maintenance.
Specifica
Specifica
| Materiale | PTFE |
| Da utilizzare con | For Any Applications Requiring Vacuum Generation and Is Ideal For Use With The Stuart Rotary Evaporator |
| Tipo di pompa | Pompa da vuoto |
| Applicazioni | For Any Applications Requiring Vacuum Generation and Is Ideal For Use With The Stuart Rotary Evaporator |
| Tensione | 230 V |