Learn More
Thermo Scientific™ Nalgene™ Syringe Filters, Non-sterile
GREENER_CHOICE
Codice prodotto. p-7235755
Click to view available options
Dimensione del poro:
0.2 μm
0.45 μm
0.8 μm
1.1 μm
Diametro (sistema metrico):
4 mm
13 mm
25 mm
Materiale (membrana):
Cellulose Acetate
Glass Fiber
Nylon
PTFE
SFCA
Dimensione della confezione:
pezzi 100
pezzi 50
Codice prodotto. 15362378
Marca: Thermo Scientific™ 7211320
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Filter sample volumes of 10 to 100mL with these nonsterile syringe filters featuring a variety of membranes and pore sizes for most laboratory applications.
Thermo Scientific™ Nalgene™ syringe filters are built to meet your lab’s clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.
Key Features
  • 10 to 100mL sample size
  • Inlet: Female Luer-Lok™
  • Outlet: Male Luer slip
  • Polypropylene housing with ID color rings
  • Pore size and membrane type printed on unit
Surfactant-free Cellulose Acetate (SFCA)
  • Low protein binding, lower extractables than standard CA
  • Good for cell culture fluids and cellular solutions
  • Polypropylene housings with blue color rings
Cellulose Acetate (CA)
  • Some protein binding and extractables
  • For sterile filtration and clarification of: buffers, aqueous solutions and reagents
  • Polypropylene housings with red color rings
Nylon
  • Naturally hydrophilic, low extractables, some protein binding
  • Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
  • Polypropylene housings with pink color rings
PTFE
  • Naturally hydrophobic
  • Excellent for HPLC/GC solvent filtration. Ideal for sterile venting of air or other gases during filtration
  • Polypropylene housings with white color rings
TRUSTED_SUSTAINABILITY
Specifica
Tipo di filtro Membrane Filter
Materiale (membrana) PTFE
Dimensione del poro 0.2 μm
Sterilità Non-sterile
Da utilizzare con (applicazione) Sterile venting of air or other gasses, sample preparation of samples for instrument analysis such as HPLC and CG
Materiale alloggiamento Polypropylene (Housing)
Quantità 100/Pk.
Tipo Filtro per siringa
Volume (sistema metrico) 2 to 10 mL
Diametro (sistema metrico) 13 mm