missing translation for 'onlineSavingsMsg'
Learn More

P2Y12/P2RY12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications

351.00€ - 676.00€

Specifica

Antigene P2Y12/P2RY12
Diluizione Western Blot - reported in scientific literature (PMID:31968618)., Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence -validated from a verified customer review, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Immunohistochemistry-Frozen -validated from a verified customer review
Applicazioni Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classificazione Polyclonal
Coniugato Unconjugated
Visualizza altre specifiche

Prodotti 2
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
18496111
Vedere i documenti
Novus Biologicals
NBP2-33870-25ul
25 μL
351.00€
25µL
Visualizzare prodotti alternativi
Risparmia fino a 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
18785033
Vedere i documenti
Novus Biologicals
NBP2-33870
0.1 mL
676.00€
0.10mL
Visualizzare prodotti alternativi
Risparmia fino a 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Descrizione

Descrizione

P2Y12/P2RY12 Polyclonal specifically detects P2Y12/P2RY12 in Human, Canine, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifica

Specifica

P2Y12/P2RY12
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
RUO
Human, Feline
Q9H244
64805
This antibody was developed against a recombinant protein corresponding to amino acids: KSFRNSLISMLKCPNSATSLSQDNRKKEQDGGDPNEETPM
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot - reported in scientific literature (PMID:31968618)., Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence -validated from a verified customer review, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Immunohistochemistry-Frozen -validated from a verified customer review
Polyclonal
Rabbit
GPCR
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
ADPG-R, Gi-coupled ADP receptor HORK3, HORK3P2Y12 platelet ADP receptor, P2T(AC), P2Y purinoceptor 12, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12ADP-glucose receptor, purinergic receptor P2RY12, purinergic receptor P2Y, G-protein coupled, 12, putative G-protein coupled receptor, SP1999G-protein coupled receptor SP1999
P2RY12
IgG
Affinity Purified
Specificity of human P2Y12/P2RY12 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen

For Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt