missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA14178
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
PA1-4178 detects human p8 (com1). PA1-4178 has been successfully used in Western blot procedures. The PA1-4178 immunogen is a synthetic peptide corresponding to the C-terminus of the human p8 protein. Maintain at 2-8oC for 3 months or at -20oC for longer storage. Acoid freeze-thaw cycles.
p8 Polyclonal antibody specifically detects p8 in Human samples. It is validated for Western Blot.
Specifica
| p8 | |
| Polyclonal | |
| Unconjugated | |
| NUPR1 | |
| 2310032H04Rik; 60B8AG; CAGA; Candidate of metastasis 1; CFAG; CGLA; COM1; CP-10; L1Ag; MA387; MIF; MRP8; NIF; Nuclear protein 1; nuclear protein 1, transcriptional regulator; nuclear protein 1-like; nuclear protein transcription regulator 1; nuclear protein, transcriptional regulator, 1; nuclear proten 1; nuclear transcriptional regulator protein 1; NUPR1; P8; protein p8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26471 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O60356 | |
| NUPR1 | |
| Synthetic peptide corresponding to the C-terminus of the human p8 protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu