Learn More
Scientific Industries SI™ Pack of 6 Metal Clip Plates to accommodate up to 3 tubes
Click to view available options
Diameter (Metric) Outer Tubing:
28 to 30 mm
Dimensione della confezione:
Pezzo
Descrizione
- Pack of 6 metal clip plates to accommodate up to 3 tubes with a diameter of 28-30mm.
- A maximum of 8 clip plates, 4 per side, may be attached to the Magnetic Platform for use with the Roto-Shake Genie, Enviro-Genie or Incubator-Genie.
- A maximum of 12 clip plates, 6 per side, may be attached to the Magnetic Platform for use with the Rotator Genie
Specifica
Specifica
| Autoclavabile | Not Autoclavable |
| Stile | Clip Plate |
| Tipi di connettore | Metal clips |
| Tipo | Metal Clip Plate |
| Materiale | Stainless Steel |
| Capacità | 3 x 28 to 30 mm Tubes/Plate |
| Da utilizzare con (apparecchiatura) | Roto-Shake Genie, Enviro-Genie, or Incubator Genie |
| Descrizione | Pack of 6 metal clip plates to accommodate up to 3 tubes |
| Diameter (English) Outer Tubing | 1.10 to 1.18 in. |
| Diameter (Metric) Outer Tubing | 28 to 30 mm |
| Vedi altri risultati |