Learn More
VACUUBRAND™ Package Rotary Vane Pump RZ 6 with Vacuum Gauge VACUU·VIEW Extended
Sfoglia Tutto Vacuubrand Prodotti
Click to view available options
Tipo di spina:
CEE
Dimensione della confezione:
Pezzo
Descrizione
- High flow rates even at vacuum levels approaching ultimate vacuum
- High water vapor tolerance due to efficient gas ballast; very good ultimate vacuum even with gas ballast
- Vacuum-tight at switch-off; external anti-suckback valve not needed
- Large oil volume: Long intervals between oil changes
- Ease of maintenance due to telescopic design
Specifica
Specifica
| Vuoto max | 0.002 mbar |
| Tipo di spina | CEE |
| Portata | 5.7 m³/hr. |
| Tipo di pompa | Rotary Vane Vacuum Pump |
| Valori nominali armadio | IP40 |
| Applicazioni | Degassing, Lyophilization, Fine Vacuum Distillation and Drying Chambers |
| Maximum Pressure | 1.1 bar |
| Tensione | 230 V |