missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pannexin-1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-82019R
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Pannexin-1 Polyclonal antibody specifically detects Pannexin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifica
| Pannexin-1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 550 | |
| MGC21309, MRS1innexin, pannexin 1, pannexin-1, PX1, UNQ2529 | |
| Rabbit polyclonal Pannexin-1 antibody was raised against a 17 amino acid peptide near the amino terminus of human Pannexin-1. The immunogen is located within amino acids 70 - 120 of Pannexin-1. Amino Acid Squence: CAVQQKNSLQSESGNLPL | |
| 0.1 mL | |
| Neuroscience | |
| 24145 | |
| Store at 4°C in the dark. | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering