Learn More
Thermo Scientific Chemicals Panobinostat
Click to view available options
Quantità:
250 mg
1 g
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
Materiale o nome chimico | Panobinostat |
Quantità | 1 g |
Peso formulazione | 349.43 |
InChI Key | FPOHNWQLNRZRFC-UHFFFAOYSA-N |
SMILES | CC1=C(CCNCC2=CC=C(C=CC(=O)NO)C=C2)C2=CC=CC=C2N1 |
IUPAC Name | N-hydroxy-3-[4-({[2-(2-methyl-1H-indol-3-yl)ethyl]amino}methyl)phenyl]prop-2-enamide |
Molecular Weight (g/mol) | 349.43 |