Learn More
Cole-Parmer™ Parker Hannifin™ PFA Compression Union Tee
Click to view available options
Dimensioni dei tubi:
1/2 in.
1/8 in.
Dimensione della confezione:
Pezzo
Descrizione
- Superior chemical resistance with durable PFA composition.
- Get a tight seal at higher pressures.
- Withstand higher pressures than barbed fittings.
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifica
Specifica
| Descrizione | Compression Union Tee |
| Autoclavabile | Autoclavable |
| Materiale | PFA |
| Dimensioni dei tubi | 1/2 in. |
| Metodo di sterilizzazione | Autoclave or Ethylene Oxide |
| Product Type | Compression Union Tee |