Learn More
Thermo Scientific™ Particle Certified HDPE Containers
Codice prodotto. 17882395
Click to view available options
Capacità (sistema metrico):
125 mL
250 mL
Certificato:
Low Particulate
Dimensione della confezione:
pezzi 24
Codice prodotto. 17882395
Marca: Thermo Scientific™ 157250W/N/LP
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Protect the quality of your products or lab samples with these reliable particle-certified HDPE containers,certified to contain < 20 particles per milliliter = 0.3 microns.
Protect the quality of your products or lab samples with reliable Thermo Scientific™ Particle-Certified HDPE Containers, tested and certified to contain <20 particles per milliliter = 0.3 microns. A Certificate of Analysis is included with each shipment.
A large variety of standard particle-certified containers are available. Nonstandard containers may be custom cleaned on request.
Key Features:
  • Narrow-mouth
  • Includes leak-proof† polypropylene caps
  • Suitable for use in sampling and packaging, including combination packaging for shipping of hazardous materials
  • Double-bagged and ready for cleanroom with no extra preparation
Recommended for:
Low particle applications
TRUSTED_SUSTAINABILITY
Specifica
Capacità 8 once, 250 ml
Capacità (sistema metrico) 250 mL
Capacità (sistema imperiale) 8 once
Colore Translucent
Diametro (sistema imperiale) 2.5 in.
Materiale HDPE
N. per custodia 24
Linea di prodotti Nalgene
Certificato Low Particulate
Volume (sistema imperiale) margine 8 oz.
Volume (sistema metrico) margine 250 mL
Certificazione/Conformità Low metals certification includes aluminum, calcium, copper, iron, potassium, magnesium, manganese, sodium, and zinc at less than 10 ppb. A certificate of analysis is included with each lot.
Closure Included
Bocca Stretta
Forma Rotondo
Altezza (sistema metrico) 133.35 mm
Product Type Water Sample Collection Bottle
Altezza (sistema imperiale) 5.25 in.
Materiale chiusura Polypropylene
Dimensione chiusura 24-415 mm
Diametro (sistema metrico) 63.5 mm
Workflow Step Upstream process solution storage. Upstream Validation. Upstream process solution storage
†The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.