missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Particle Certified HDPE Containers

Codice prodotto. p-4521652
Click to view available options
Capacità (sistema metrico):
125 mL
250 mL
Certificato:
Low Particulate
Dimensione della confezione:
pezzi 24
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 17872395

Marca: Thermo Scientific™ 156125W/N/LP

Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Protect the quality of your products or lab samples with these reliable particle-certified HDPE containers,certified to contain < 20 particles per milliliter = 0.3 microns.

Protect the quality of your products or lab samples with reliable Thermo Scientific™ Particle-Certified HDPE Containers, tested and certified to contain <20 particles per milliliter = 0.3 microns. A Certificate of Analysis is included with each shipment.

A large variety of standard particle-certified containers are available. Nonstandard containers may be custom cleaned on request.

Key Features:

  • Narrow-mouth
  • Includes leak-proof† polypropylene caps
  • Suitable for use in sampling and packaging, including combination packaging for shipping of hazardous materials
  • Double-bagged and ready for cleanroom with no extra preparation

Recommended for:

Low particle applications

TRUSTED_SUSTAINABILITY

Specifica

Capacità 4 oz., 125 mL
Capacità (sistema metrico) 125 mL
Capacità (sistema imperiale) 4 oz.
Colore Translucent
Diametro (sistema imperiale) 2 in.
Materiale HDPE
N. per custodia 24
Linea di prodotti Nalgene
Certificato Low Particulate
Volume (sistema imperiale) margine 4 oz.
Volume (sistema metrico) margine 125 mL
Certificazione/Conformità Low metals certification includes aluminum, calcium, copper, iron, potassium, magnesium, manganese, sodium, and zinc at less than 10 ppb. A certificate of analysis is included with each lot.
Closure Included
Bocca Stretta
Forma Rotondo
Altezza (sistema metrico) 101.6 mm
Product Type Water Sample Collection Bottle
Altezza (sistema imperiale) 4 in.
Materiale chiusura Polypropylene
Dimensione chiusura 24-415 mm
Diametro (sistema metrico) 50.8 mm
Workflow Step Upstream process solution storage. Upstream Validation. Upstream process solution storage
Vedi altri risultati Mostra meno risultati

†The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato