missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ U2OS PTGER1 β-Arrestin Cell Line

Product Code. 17394039
Encompass
Click to view available options
Quantità:
2 Vials
This item is not returnable. View return policy

Product Code. 17394039

Brand: DiscoverX 930937C3

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

The PathHunter™ U2OS PTGER1 β-Arrestin Cell Line measures PTGER1 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells.

The PathHunter™ PTGER1 β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin E2) based activation of PTGER1 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged PTGER1 and EA-tagged β-Arrestin 2. Activation of PTGER1 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry β-Arrestin Recruitment
Linea cellulare U2OS
Da utilizzare con (applicazione) Cell Lines
Primary Agonist Prostaglandin E2
Quantità 2 Vials
Target Entry PTGER1
Numero di accesso NM_000955.2
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.