Learn More
DiscoverX pCMV-ARMS2-ProLinkTM 2 Vector
Click to view available options
Quantità:
1 Vector
Dimensione della confezione:
1 set
Descrizione
The pCMV-ARMS2-ProLinkTM 2 Vector is a mammalian cloning vector for use in expressing a protein of interest fused to the ProLink 2 (PK 2) tag at the C-terminus. The PK tag is a version of ED used for EFC assays (in combination with EA). The ProLink 2 tag has a higher affinity for EA, making it ideal for use with weaker protein:protein interactions.The ARMS (Arrestin Recruitment Modifying Sequence) PK tags contain GRK phosphorylation sites to improve ligand-dependent arrestin recruitment to GPCRs and improve assay performance. Provided as a vial of 10μg vector in 20μL liquid buffer.
Specifica
Specifica
| Product Type | Cloning Vector |
| Quantità | 1 Vector |
| Da utilizzare con (applicazione) | Cloning Vectors |