Learn More
DiscoverX pCMV-ProLabel™ N Vector
Click to view available options
Quantità:
1 Vector
Descrizione
The pCMV-ProLabel™ N Vector is a mammalian cloning vector for use in expressing a protein of interest fused to the ProLabel (PL) tag at the N-terminus. The PL tag is a version of ED used for EFC assays (in combination with EA) where ED and EA are contained in separate cellular compartments. Provided as a vial of 10μg vector in 20μL liquid buffer.
Specifica
Specifica
| Product Type | Cloning Vector |
| Quantità | 1 Vector |
| Da utilizzare con (applicazione) | Cloning Vectors |