missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDK4 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-07049UV
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
PDK4 Polyclonal specifically detects PDK4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifica
| PDK4 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 4, mitochondrial, EC 2.7.11, EC 2.7.11.2, FLJ40832, Pyruvate dehydrogenase kinase isoform 4, pyruvate dehydrogenase kinase, isoenzyme 4, pyruvate dehydrogenase kinase, isozyme 4, pyruvate dehydrogenase, lipoamide, kinase isozyme 4, mitochondrial | |
| Rabbit | |
| Affinity Purified | |
| Cancer, Lipid and Metabolism, Mitochondrial Markers, Protein Kinase, Signal Transduction | |
| 5166.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| DyLight 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PDK4 | |
| Synthetic peptide made to an internal portion of the human PDK4 protein (within residues 200-250). [Swiss-ProtNo. Q16654] | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido