Learn More
ACROBiosystems PE-Labeled Human SLAMF7 / CRACC / CD319 Protein, His Tag
Marca: ACROBiosystems SL7-HP2H3-25tests
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
SLAM family member 7 (SLAMF7) is also known as CD2-like receptor-activating cytotoxic cells (CRACC), Membrane protein FOAP-12, CD antigen CD319, Novel Ly9, Protein 19A, which is a single-pass type I membrane protein and a member of the CD2 family of cell surface receptors. SLAMF7 is expressed in spleen, lymph node, peripheral blood leukocytes, bone marrow, small intestine, stomach, appendix, lung and trachea. Isoform 1 of SLAMF7 mediates NK cell activation through a SH2D1A-independent extracellular signal-regulated ERK-mediated pathway. May play a role in lymphocyte adhesion. Isoform 3 of SLAMF7 does not mediate any NK cell activation.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.6 kDa.Specifica
AAH27867 | |
25 tests | |
Ser 23 - Met 226 | |
SLAMF7,CD319,CS1,CRACC,19 A,FOAP-12 | |
PE Labeled | |
His Tag | |
HEK293 | |
Fluorescent Labeled Protein |
25.6 kDa | |
PE-Labeled Human SLAMF7, His Tag (SL7-HP2H3) is produced via conjugation of PE to Human SLAMF7, His Tag under optimal conditions. Human SLAMF7, His Tag is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Met 226 (Accession # Q9NQ25-1). | |
-20°C, aviod light | |
SLAMF7 | |
Human | |
PBS, 0.5% BSA, pH7.4 | |
Powder |