Learn More
Thermo Scientific Chemicals Pectinase, Aspergillus niger
Codice prodotto. p-7076612
Click to view available options
Quantità:
250 mg
1 g
Dimensione della confezione:
1g
250mg
Codice prodotto. 15455659
Marca: Thermo Scientific Chemicals J63408.MD
Richiesta di formato massivo o personalizzato
Edge
Used in plant protoplast preparation to digest cell wall prior to organelle isolation. Pectinase helps facilitate the maceration of plant materials and extraction processes including organelle preparation.
Pectinase, Aspergillus niger is used in plant protoplast preparation to digest cell wall prior to organelle isolation. Pectinase helps facilitate the maceration of plant materials and extraction processes including organelle preparation. It is used in the enzyme preparation. Used mostly to reduce viscosity, to improve filtration to clarify products, to avoid particle sedimentation and to prevent pectin gel formation in the manufacture of fruit juice and wine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifica
Quantità 250 mg
Forma Powder