missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical PEEK Tubing, 1/32 in. O.D., 100 ft

Codice prodotto. 15770979
Click to view available options
Diametro (sistema imperiale) interno:
0.0025 in.
0.004 in.
0.005 in.
0.007 in.
0.010 in.
0.015 in.
0.020 in.
Colore:
Black
Blue
Gray
Natural
Orange
Red
Yellow
Dimensione della confezione:
Pezzo
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 15770979

Marca: Trajan Scientific and Medical 1301005001100F

Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Designed specifically for modern LC, LCMS and automation useage. A flexible alternative to stainless steel tubing in high pressure applications, it ensures reproducibility in your chromatography flow rates and results through smooth internal surface along with consistent and tight toleranced IDs

  • Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
  • Bio-inert ad bio-compatible
  • Color coded to industry standards for easy identification of inner diameter
  • Leach-free colorants
  • Smooth inner walls and precise IDs
  • Works well with either polymer fittings as well as stainless steel fittings
  • Can be formed to specific customer needs

Specifications

Colore Red
Intervallo di pH 2 to 14
Diametro (sistema imperiale) interno 0.005 in.
Diametro (sistema imperiale) esterno 1/32 in.
Overall Length (English) 100 ft.
Materiale PEEK
Pressione massima 5000 psi (345 bar)
Temperatura operativa max (sistema metrico) 100°C

Solo per ricerca

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.