missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PEI STAR-Go™ transfection reagent
Click to view available options
Quantità:
5 mL
100 mL
Dimensione della confezione:
100mL
5mL
Descrizione
PEI STAR-Go™ is a ready-to-use aqueous formulation of our high performance PEI STAR™ transfection reagent (Cat. No. 7854), provided as a sterile-filtered solution. PEI STAR™ is a chemically-defined, linear polyethylenimine (PEI) transfection reagent for affordable and scalable transient gene expression. PEI is a synthetic polymer with an exceptionally high positive charge density in pH-neutral solutions. Positively charged PEI binds strongly to negatively charged DNA and imparts a net cationic charge, allowing the DNA to enter cells. PEI is a non-viral vector commonly used to transfect HEK293, CHO, and a range of other eukaryotic cell lines. Applications include production of lentivirus and rAAV, recombinant proteins, and antibodies. Supplied as a 1mg/mL aqueous solution
PEI STAR-Go is a trademark of Bio-Techne Corp.
Specifica
Specifica
| Materiale o nome chimico | Poly[imino(1,2-ethanediyl)] hydrochloride |
| CAS | 49553-93-7 |
| Quantità | 100 mL |
| Bersaglio | Transfection Reagent |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto