Learn More
3M™ PELTOR™ Optime™ II Earmuffs
Click to view available options
Modello:
H520A-472-GB
H520F-409-GQ
H520F-460-GB
H520P3A-410-GQ
H520P3AF-410-GQ
H520P3B-410-GQ
H520P3E-410-GQ-01
H520P3E-467-GB
H520P3EA-410-GQ
H520P3F-410-GQ
H520P3G-410-GQ
H520P3H-410-GQ
H520P3H-467-GB
H520P3K-410-GQ
Dimensione della confezione:
pezzi 10
pezzi 20
pezzi 6
Descrizione
- Ideal hearing protection against noise arising from environments with substantial industrial noise or construction machinery, like airports and agricultural work
- Soft wide cushions helps reduce pressure around the ears and improves comfort and wearability
- Large space inside cup helps reduce moisture and heat build-up
- Easy to replace cushions and inserts helps keep them hygienically clean
Specifica
Specifica
| Colore | Green |
| Certificazione/Conformità | CE Approved, EN 352-3: 2002 |
| Modello | H520P3F-410-GQ |
| Noise Reduction Rating (NRR) | 30 dB |
| Product Type | Ear Muffs |
| Dielectric | No |