Learn More
3M™ 3M™ PELTOR™ Cuffie WS™ ALERT™ X, giallo brillante, bardatura temporale, compatibili con l'app mobile, MRX21A4WS6
Codice prodotto. 17854931
Click to view available options
Stile:
Headband
Helmet Mounted
Modello:
MRX21A4WS6
MRX21P3E4WS6
Dimensione della confezione:
pezzi 10
Codice prodotto. 17854931
Marca: 3M™ 7100256658
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Le cuffie con tecnologia Bluetooth™ e microfono a cancellazione del rumore si collegano al cellulare per telefonare a mani libere e per lo streaming in ambienti rumorosi.
  • Attenuating hearing protector comes in two models: headband or a safety helmet attachment
  • Noise-cancelling boom microphone for clear speech transmission in noisy environments
  • Level-dependent function for ambient listening to help promote auditory situational awareness
  • Connection to an iOS and Android compatible mobile app, 3MTM Connected Equipment, to more easily set-up and adjust the headset. The app is available to download from App Store and Google Play.
  • Mobile app allows you to receive reminders when to change replacement cushions (hygiene kit) and choose the frequency of these alerts
  • Push-to-Listen feature to easily and instantaneously listen to your surroundings
  • Voice guided feedback after setting adjustment. No need to remove the headset for any adjustments
  • The headset will automatically turn off after four hours of non-use to save battery life
  • Replaceable cushions (hygiene-kit) and covers
TRUSTED_SUSTAINABILITY
Specifica
Modello MRX21P3E4WS6
Noise Reduction Rating (NRR) 30 dB
Stile Helmet Mounted
Dettagli comunicazione Compatible with Mobile App