Learn More
3M™ 3M™ PELTOR™ Cuffie WS™ ALERT™ X, giallo brillante, bardatura temporale, compatibili con l'app mobile, MRX21A4WS6
Click to view available options
Stile:
Headband
Helmet Mounted
Modello:
MRX21A4WS6
MRX21P3E4WS6
Dimensione della confezione:
pezzi 10
Descrizione
- Attenuating hearing protector comes in two models: headband or a safety helmet attachment
- Noise-cancelling boom microphone for clear speech transmission in noisy environments
- Level-dependent function for ambient listening to help promote auditory situational awareness
- Connection to an iOS and Android compatible mobile app, 3MTM Connected Equipment, to more easily set-up and adjust the headset. The app is available to download from App Store and Google Play.
- Mobile app allows you to receive reminders when to change replacement cushions (hygiene kit) and choose the frequency of these alerts
- Push-to-Listen feature to easily and instantaneously listen to your surroundings
- Voice guided feedback after setting adjustment. No need to remove the headset for any adjustments
- The headset will automatically turn off after four hours of non-use to save battery life
- Replaceable cushions (hygiene-kit) and covers
Specifica
Specifica
| Modello | MRX21A4WS6 |
| Noise Reduction Rating (NRR) | 30 dB |
| Stile | Headband |
| Dettagli comunicazione | Compatible with Mobile App |