Learn More
Honeywell™ PERFECT POLY™ Gray Gloves
Click to view available options
Size:
6
7
8
9
Dimensione della confezione:
1 paio
Descrizione
- Gray polyamide knitted glove
- Grey polyurethane coating on the palm
- The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
- The glove offers maximum comfort and added sturdiness
- The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The ventilated back and PU coating optimise the glove's breathability
- Polyamide with its polyurethane coating provide excellent abrasion and tear resistance
- The PU coating ensures a good grip
- Non pilling, the polyamide is suitable for precision work
- The gloves are certified as silicone-free to ensure their use in painting
- Dark color makes it more soilproof, thus ensuring a longer life
Specifica
Specifica
| Certificazione/Conformità | EN 420-2003 Basic standard, EN 388-2003 Mechanical risks |
| Descrizione | 15 gauge |
| Colore | Gray |
| Materiale | Polyamide, Polyurethane |
| Size | 6 |
| Lunghezza (sistema metrico) | 25 cm |