Learn More
Honeywell™ PERFECT POLY™ Skin Gloves
Codice prodotto. p-7159548
Click to view available options
Size:
7
8
9
Dimensione della confezione:
1 paio
Codice prodotto. 15836795
Marca: Honeywell™ 240026007
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Honeywell™ PERFECT POLY™ Skin Gloves for handling fine products in a dry or slightly greasy or soiled environment.
  • Polyamide knitted glove
  • Gray polyurethane coating on palm and tips of fingers
  • Elastic wrist
  • The knitting technique helps to obtain a glove that conforms snugly to the hand as well as outstanding dexterity
  • The glove offers maximum comfort and added sturdiness
  • The ultra tight weave gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The ventilated back and ultra light coating optimise the glove's breathability
  • Polyamide with its polyurethane coating provide excellent abrasion and tear resistance
  • Non pilling, the polyamide is suitable for precision work
  • The gloves are certified as silicone-free to ensure their use in painting
  • Blue color and gray coating make the glove more soilproof, thus ensuring a longer life
TRUSTED_SUSTAINABILITY
Specifica
Certificazione/Conformità EN 420-2003 Basic standads, EN 388-2016 Mechanical hazards
Descrizione 18 gauge
Colore Blue, Gray
Materiale Polyamide, Polyurethane
Size 7
Lunghezza (sistema metrico) 24 cm