Learn More
Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA) Acylation Reagents
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Componenti standard:
Heptafluoro-butanoic acid anhydride
Pentafluoro-propanoic acid anhydride
Trifluoroacetic acid anhydride
Quantità:
25 g
100 g
10 x 1 mL
Dimensione della confezione:
10mL
Pezzo
pezzi 10
Descrizione
Prepare fluoracyl derivatives for GC/MS and for flame ionization and electron-capture detection techniques with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA). These reagents react readily with alcohols, phenols, and amines.
- Used to prepare fluoracyl derivatives for GC/MS
- Produce stable volatile derivatives for FID and ECD techniques
Specifica
Specifica
| Da utilizzare con | GC/MS |
| Rischi | Corrosive |
| Conservazione | Room Temperature |
| Quantità | 10 x 1 mL |
| Componenti standard | Pentafluoro-propanoic acid anhydride |
| Concentrazione standard | 98 to 100% |