Learn More
Thermo Scientific™ Perfluoroacylimidazoles (TFAI and HFBI) Acylation Reagents
Codice prodotto. p-4527519 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Componenti standard:
TFAI
Dimensione della confezione:
10mL
Codice prodotto. 11747067
Marca: Thermo Scientific™ TS48882
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Conduct acylation of hydroxyl groups and primary and secondary amines with the Thermo Scientific™ Perfluoroacylimidazoles (TFAI and HFBI).
Conduct acylation of hydroxyl groups and primary and secondary amines with the Thermo Scientific™ Perfluoroacylimidazoles (TFAI and HFBI). Compared to acid chlorides and anhydrides, the reactions of fluorinated acylimidazoles are smooth and quantitative, and produce no acid byproducts. The principal byproduct is imidazole, which is relatively inert. The volatile and stable fluoroacyl derivatives are ideal for FID and electron-capture detection (ECD) techniques.
Applications
Used in bifunctional derivatization schemes and in exchange reactions in which TMS derivatives are converted to HFB derivatives.
Hydroxyl groups of catecholamines are derivatized with TMSI, followed by conversion of the amines to acylamides with HFBI.
Tryptamine and metabolites present in spinal fluid have been analyzed by ECD using HFBI.
TRUSTED_SUSTAINABILITY
Specifica
Da utilizzare con FID, ECD
Rischi Corrosive, Flammable
Conservazione 2°C to 8°C
Quantità 10 x 1 mL Ampules
Componenti standard TFAI
Concentrazione standard 98 to 100%
Descrizione articolo TFAI; 10 x 1 mL ampules