Learn More
Perfluorotetradecanoic acid, 96%
Click to view available options
Quantità:
1 g
5 g
Dimensione della confezione:
1g
5g
Descrizione
Perfluorotetradecanoic acid is used for GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Perfluorotetradecanoic acid is used for GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.
Solubility
It is insoluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Perfluorotetradecanoic acid is used for GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.
Solubility
It is insoluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifica
Specifica
| Materiale o nome chimico | Perfluorotetradecanoic acid |
| Punto di fusione | 130°C to 132°C |
| Punti di ebollizione | 192°C (60 mmHg) |
| Quantità | 1 g |
| Numero UN | UN3261 |
| Beilstein | 1811438 |
| Informazioni di solubilità | It is insoluble in water. |
| Peso formulazione | 714.12 |
| Percent Purity | 96% |