Learn More
Labcon™ PerformR™ 12x75mm Round Bottom Flow Cytometry Tubes
GREENER_CHOICE
Codice prodotto. p-200112170 Sfoglia Tutto Labcon Prodotti
Click to view available options
Colore:
Blue
Green
Dimensione della confezione:
pezzi 1000
Codice prodotto. 30280329
Marca: Labcon™ 33503670009
Polypropylene tubes for cell sorting and general flow cytometry use
  • Flow Cytometry Tubes
  • ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Polypropylene USP Class VI resin
  • Centrifuging to 3,000 x G
  • Freezable to -80C
  • Autoclavable
  • Capacity: 5 mL
  • Seal with plug or dual position caps sold separately
  • 12 mm W. 75 mm H.
  • Made in USA
Specifica
Certificazione/Conformità ISO 9001:2015, FDA CFR 21 GMP
Colore Green
Product Type Flow Cytometer Accessory