Learn More
IKA™ PETTE MicroKit B
Sfoglia Tutto IKA Prodotti
Click to view available options
Volume (sistema metrico):
Variable
Unit Size:
Pezzo
Description
- The package consists of three single channel pipettes with variable volumes in the micro-volume range, a complimentary stand that holds up to three pipettes and two pipette tip boxes. This makes it a convenient starter package for researchers.
- Features:
- Mechanical air-cushion pipettes manufactured in Germany
- Fully automated calibration by robots
- Volume-locking feature to prevent any accidental volume changes
- Large and clearly visible display
- Ergonomic and lightweight pipette with low pipetting and tip ejection forces to reduce strain and fatigue
- Durable and robust construction using high quality UV resistant and chemical resistant materials
- Fully autoclavable to ensure sterility and compliance to stringent laboratory standards
- Maximum user convenience with tool-less disassembly and calibration adjustment for regular maintenance
Specifications
Specifications
| Descrizione | Air Displacement Piston-Operated Pipette |
| Autoclavabile | Autoclavable |
| Numero di canali | 1 |
| Product Line | PETTE |
| Sterilità | Sterile |
| Tipo | Pipette Package |
| Volume (sistema metrico) | Variable |
| Include | Three Single Channel Pipettes With Variable Volumes In The Micro-Volume Range, One Complimentary Stand That Holds Up To Three Pipettes, And Two Pipette Tip Boxes |