missing translation for 'onlineSavingsMsg'
Learn More

PGC1 alpha Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Novus Biologicals NBP1-04676AF350

Codice prodotto. 18223633

  • 843.00€ / 0.10mL

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Questo articolo non è restituibile. Consulta la politica di reso

Beskrivelse

Beskrivelse

PGC1 alpha Polyclonal antibody specifically detects PGC1 alpha in Human, Mouse, Rat, Porcine, Goat, Hamster, Sheep, Squirrel samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow (Intracellular), KnockDown
TRUSTED_SUSTAINABILITY
Tekniske data

Tekniske data

PGC1 alpha
Polyclonal
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), Knockout Validated
LEM6, Ligand effect modulator 6, ligand effect modulator-6, L-PGC-1alpha, peroxisome proliferative activated receptor, gamma, coactivator 1, alpha, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-3ext, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-8a, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5-NT, peroxisome proliferator-activated receptor gamma coactivator 1-alpha, peroxisome proliferator-activated receptor gamma, coactivator 1 alpha, PGC1, PGC-1(alpha), PGC1A, PGC-1-alpha, PGC1APGC-1(alpha), PGC1peroxisome proliferative activated receptor, gamma, coactivator 1, PGC-1v, PPAR gamma coactivator variant form, PPAR gamma coactivator-1, PPARgamma coactivator 1alpha, PPAR-gamma coactivator 1-alpha, PPARGC1, PPARGC-1-alpha
This PGC1 alpha Antibody was developed against a recombinant protein made to an internal portion of the human PGC-1 alpha protein (within residues 400-550). [Swiss-Prot# Q9UBK2].
0.1 mL
Alzheimers Research, Cancer, Cardiovascular Biology, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Epigenetics, Lipid and Metabolism, Mitochondrial Mediated Pathway, mTOR Pathway, Neuroscience, Transcription Factors and Regulators
10891
Store at 4°C in the dark.
IgG
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow Cytometry, KnockDown
Alexa Fluor 350
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
Primary
Human, Mouse, Rat, Pig, Goat, Hamster, Sheep, Squirrel
Purified
Produktforslag

Produktforslag

Videoer
SDS
Dokumenter

Dokumenter

Produktcertifikater
Kampagner

Kampagner

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.