Learn More
pH Singles Calibration Solutions in 20mL Pouches, Oakton™
Click to view available options
pH:
10.00
Dimensione della confezione:
20mL
Descrizione
The pH Singles are ideal for field calibration—just select the pouches needed and carry them to your test site for on-the-spot calibration and verification. No bottles to carry or clean. Pouches are air-tight for maximum freshness and unquestionable reliability. Packets are labeled with temperature versus pH reference tables. Sold in packs of twenty 20mL pouches.
Certifications
NIST traceable, All solutions are certified traceable to NIST™ standards.
Specifica
Specifica
| pH | 10.00 |
| Quantità | 20 mL Pouches |