Learn More
PHACTR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 23446-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
PHACTR1 plays a role in cell motility and in the formation of tubules by endothelial cells. PHACTR1 also binds actin monomers (G actin) and plays a role in the reorganization of the actin cytoskeleton and in formation of actin stress fibers. Further, PHACTR1 regulates PPP1CA activity and is required for normal cell survival.Specifica
| PHACTR1 | |
| Polyclonal | |
| Unconjugated | |
| PHACTR1 | |
| KIAA1733, PHACTR1, RPEL, RPEL1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 218194, 221692, 306844 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62024, Q2M3X8, Q9C0D0 | |
| Phactr1 | |
| PHACTR1 Fusion Protein Ag20125 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |