Learn More
Applied Biosystems™ Pharma KingFisher™ Flex 96 Standard Plates
Descrizione
These standard plates are for use with the Pharma KingFisher Flex 96 Deep-Well Head Magnetic Particle Processor or MagMAX Express-96 Standard Magnetic Particle Processor in combination with PrepSEQ ResDNA kits and ProteinSEQ kits. The standard configuration differs from the deep-well configuration in that the volume of sample input is from 20–200 μL per well compared to 100–1000 μL per well for the deep-well configuration.
Optimize performance using KingFisher Flex System consumables designed specifically for use with these instruments. KingFisher Flex disposable plastics (tip combs and microplates) are made of polypropylene and are ideal for magnetic particle processing because of their low binding affinity for biomolecules. Their special design enables excellent recovery of magnetic beads.
Specifica
Specifica
| Codice a barre | No Barcode |
| Content And Storage | 48 standard-well 96-well plates |
| Imballaggio | 48 Plates |
| Product Line | KingFisher Flex |
| Colore | Clear |
| Product Type | Microplate |
| Da utilizzare con (apparecchiatura) | KingFisher™ Flex |
| Materiale | Polypropylene |
| Volume (sistema metrico) pozzetto | 1000 μL |
| No. of Wells | 96 |
| Vedi altri risultati |
For Research Use Only. Not for use in diagnostic procedures.
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.