Learn More
Masterflex™ Tubi flessibili Pharmed BPT
Codice prodotto. 15134663
Click to view available options
Diametro (sistema metrico) interno:
4.8 mm
Dimensione della confezione:
25ft
Codice prodotto. 15134663
Marca: Masterflex™ 9688008
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Use this tubing for peristaltic pumps and sensitive cell cultures. Saint Gobain Performance Plastics™ PharMed™ BPT Hoses are less permeable to gases and vapors than silicone tubing and are ideal for fermentation, separation, purification and process monitoring and control systems.
  • Outlasts silicone tubing in peristaltic pumps by up to 30 times
  • Low particulate spallation
  • Can be autoclaved repeatedly
  • Withstands repeated CIP and SIP cleaning and sterilization
  • Fully tested to ISO 10993 standards to facilitate validation process
  • Meets USP Class VI, FDA and NSF criteria
Typical Applications
  • Diagnostic test product manufacturing
  • Cell harvest and media process systems
  • Vaccine manufacturing
  • Bioreactor process lines
  • Production filtration and fermentation
  • Sterile filling
  • Shear-sensitive fluid transfer
Specifica
Certificazione/Conformità FDA ( 21 CFR 177.2600 ), NSF (standard 51 ), USP 23 Classe VI
Autoclavabile Autoclavable
Diametro (sistema metrico) interno 4.8 mm
Diametro (sistema metrico) esterno 8.0 mm
Overall Length (Metric) 7.5 m
Spessore (sistema metrico) 1.6 mm
Da utilizzare con (applicazione) Industria farmaceutica, biotecnologie, trasferimenti di colture cellulari, cellule o terreni fotosensibili
Min. Operating Temperature (Metric) °
Quantità confezione 1 Each
Sterilità Sterili
Colore Opaco
Flexibility Flessibile
Max. Operating Temperature (Metric) da –50 a 135 °C