Learn More
Masterflex™ L/S™ PharMed™ BPT Spooled Pump Tubing
Codice prodotto. 11735228
Click to view available options
Dimensioni dei tubi:
14
15
16
17
18
24
35
36
Dimensione della confezione:
Pezzo
Codice prodotto. 11735228
Marca: Masterflex™ 9568717
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Use this spooled tubing for cell and tissue culture work and pharmaceutical uses. Masterflex™ L/S™ PharMed™ BPT Spooled Pump Tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport.
  • Opaque beige spooled pump tubing requires less storage space than the equivalent amount of individual tubing coils
  • Heat-sealable, bondable and formable
  • Ideal for light-sensitive samples
  • Nontoxic, nonhemolytic, extremely low permeability
  • Resists acids, alkalis, oxidizing agents and animal and vegetable oils
  • Reduce scrap, cut only as much as you need
  • Ensure that you always have enough tubing on hand
Nearly any Masterflex pump tubing formulation can be spooled. Call our Application Specialists for a quote or to place a special order
Specifica
Certificazione/Conformità FDA approved; Prop 65 NA (no Prop 65 chemicals)
Autoclavabile Autoclavable
Diametro (sistema metrico) interno 6.4 mm
Overall Length (English) 500 ft.
Overall Length (Metric) 152.4 m
Min. Operating Temperature (Metric) -20°C
Dimensioni dei tubi 17
Portata 1700 mL/min
Flessibile flangiato 1/4 in.
Metodo di sterilizzazione Autoclave, Ethylene Oxide, Gamma Irradiation
Colore Beige
Pressione massima 20 psi (1.4 bar)
Livello di vuoto 510 mm Hg (20 in. Hg)
Max. Operating Temperature (Metric) 100°C
Product Type Pump Tubing