Learn More
Motic™ Phase Slider, Empty
Click to view available options
Tipo:
Phase Slider, Empty
Dimensione della confezione:
Pezzo
Descrizione
- Boost contrast and detail discrimination without staining or fixing, thus permitting live cell study.
- Must be matched and aligned to the objective phase plate.
Specifica
Specifica
| Tipo | Phase Slider, Empty |