Learn More
DiscoverX Phenylephrine
Click to view available options
:
10 mg
Description
Phenylephrine hydrochloride, or NeoSynephrine, is a synthetic sympathomimetic drug compound structurally similar to epinephrine that is a selective agonist of the α1-adrenergic receptor on blood vessels, causing vasoconstriciton. Its in-vivo effects include decongestion, hypertension, and pupil dilation. It is a common ingredient in many decongestants, and for cold, flu, allergy, or breathing illnesses. It acts as an agonist of ADRA1A, B, and C. Provided as 10mg of dry powder.
Specifications
Specifications
| Quantità | 10 mg |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.