Learn More
Invitrogen™ Phospho-4EBP1 (Thr37, Thr46) Recombinant Rabbit Monoclonal Antibody (4EB1T37T46-A5), APC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA536938
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This gene encodes one member of a family of translation repressor proteins. The protein directly interacts with eukaryotic translation initiation factor 4E, which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5' end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.
Specifica
| Phospho-4EBP1 (Thr37, Thr46) | |
| Recombinant Monoclonal | |
| APC | |
| EIF4EBP1 | |
| 4EBP1; 4E-BP1; AA959816; BP-1; eIF4E-binding protein 1; EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1; eukaryotic translation initiation factor 4E-binding protein 1; MGC431; MGC4316; P/OKCL.6; PHAS-1; PHAS-I; Phosphorylated heat- and acid-stable protein regulated by insulin 1 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 116636, 13685, 1978 | |
| 4°C | |
| Liquid |
| Flow Cytometry | |
| 4EB1T37T46-A5 | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| Q13541, Q60876, Q62622 | |
| EIF4EBP1 | |
| A synthetic phospho-peptide corresponding to residues surrounding Thr37/46 of human phospho 4E-BP1. | |
| 100 Tests | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG κ |