missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-KCC2 (Ser940) Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA595678
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Antigen Affinity Purified from Pooled Serum.
Mediates electroneutral potassium-chloride cotransport in mature neurons. Transport occurs under isotonic conditions, but is activated 20-fold by cell swelling. Important for Cl(-) homeostasis in neurons.
Specifica
| Phospho-KCC2 (Ser940) | |
| Polyclonal | |
| Unconjugated | |
| SLC12A5 | |
| EIEE34; EIG14; Electroneutral potassium-chloride cotransporter 2; erythroid K-Cl cotransporter 2; furosemide-sensitive K-Cl cotransporter; hKCC2; KCC 2; Kcc2; KCC2a variant 1; KCC2a-S25 variant 1; KCC2b variant 2; K-Cl cotransporter 2; KIAA1176; mKCC2; mKIAA1176; neuronal K-Cl cotransporter; neuronal-specific K-Cl cotransporter; rKCC2; S12A5; SLC12A5; solute carrier family 12 (potassium/chloride transporter), member 5; solute carrier family 12 (potassium-chloride transporter), member 5; solute carrier family 12 member 5; solute carrier family 12, (potassium-chloride transporter) member 5; solute carrier family 12, member 5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 171373, 57138 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| 10mM HEPES with 100 μg/mL BSA, 0.15M NaCl, 50% glycerol and no preservative, pH 7.5 | |
| Q63633, Q91V14 | |
| SLC12A5 | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser940 conjugated to KLH. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto